Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Doxorubicin hydrochloride, 10 mg/ml in distilled water, sterile filtered

Catalog No. AAJ67364XF
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67364XF 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67364XF Supplier Thermo Scientific Chemicals Supplier No. J67364XF

CAS: 25316-40-9 | C27H30ClNO11 | 579.98 g/mol

It acts as an antitumor antibiotic agent that inhibits DNA topoisomerase II and acts as a DNA intercalator that inhibits nucleic acid synthesis and induces apoptosis. It reduces intracellular tau levels. By intercalating within DNA, doxorubicin inhibits nucleic acid synthesis and induces apoptosis by inducing the accumulation of the p53 tumor suppressor protein. It is an is an inhibitor of AMPK, TERT and POLR. its fluorescent property has been exploited for the measurement of drug efflux pump activities as well as resolving the important question of intracellular localization of various multidrug resistance proteins and the role of subcellular organelles (Golgi and lysosome) in the sequestration of drugs and its implication in drug resistant phenotypes. It is applied as an antineoplastic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It acts as an antitumor antibiotic agent that inhibits DNA topoisomerase II and acts as a DNA intercalator that inhibits nucleic acid synthesis and induces apoptosis. It reduces intracellular tau levels. By intercalating within DNA,
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25316-40-9
Molecular Formula C27H30ClNO11
Molecular Weight (g/mol) 579.98
MDL Number MFCD00077757,MFCD00077757,MFCD00941448
InChI Key MWWSFMDVAYGXBV-FGBJBKNOSA-N
PubChem CID 129626538
IUPAC Name hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride
SMILES [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O

Specifications

Type Doxorubicin Hydrochloride
Physical Form Liquid
Quantity 1 mL
Solubility Information Fully miscible in water and soluble in 50 mM in DMSO,methanol and isotonic sodium chloride (>10mg/ml). Insoluble in chloroform,and ether.
Chemical Name or Material Doxorubicin hydrochloride, 10mg/mL in distilled water, sterile-filtered
Hazard Category H319-H340-H350-H361
Hazard Statement GHS H Statement
H350
May cause cancer.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
EINECSNumber 246-818-3
RTECSNumber QI9295900
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.