CAS RN 25316-40-9
Doxorubicin hydrochloride
CAS: 25316-40-9 Molecular Formula: C27H30ClNO11 Molecular Weight (g/mol): 579.98 MDL Number: MFCD00077757,MFCD00077757,MFCD00941448 InChI Key: MWWSFMDVAYGXBV-FGBJBKNOSA-N Synonym: doxorubicin hydrochloride,adriamycin,doxorubicin hcl,adriacin,adriblastina,rubex,adriamycin hydrochloride,adriblastin,adriamycin pfs PubChem CID: 129626538 IUPAC Name: hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride SMILES: [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O
Doxorubicin Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
Doxorubicin hydrochloride, MP Biomedicals™
CAS: 25316-40-9 Molecular Formula: C27H30ClNO11 Molecular Weight (g/mol): 579.98 MDL Number: MFCD00077757,MFCD00077757,MFCD00941448 InChI Key: MWWSFMDVAYGXBV-FGBJBKNOSA-N Synonym: doxorubicin hydrochloride,adriamycin,doxorubicin hcl,adriacin,adriblastina,rubex,adriamycin hydrochloride,adriblastin,adriamycin pfs PubChem CID: 129626538 IUPAC Name: hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride SMILES: [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O
Doxorubicin Hydrochloride 95.0+%, TCI America™
CAS: 25316-40-9 Molecular Formula: C27H30ClNO11 Molecular Weight (g/mol): 579.98 MDL Number: MFCD00077757,MFCD00077757,MFCD00941448 InChI Key: MWWSFMDVAYGXBV-FGBJBKNOSA-N Synonym: doxorubicin hydrochloride,adriamycin,doxorubicin hcl,adriacin,adriblastina,rubex,adriamycin hydrochloride,adriblastin,adriamycin pfs PubChem CID: 129626538 IUPAC Name: hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride SMILES: [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O
R&D Systems™ Doxorubicin hydrochloride
CAS: 25316-40-9 Molecular Formula: C27H30ClNO11
Fisher Scientific Doxorubicin Hydrochloride, Fisher BioReagents
CAS: 25316-40-9 Molecular Formula: C27H30ClNO11 Molecular Weight (g/mol): 579.98 MDL Number: MFCD00077757,MFCD00077757,MFCD00941448 InChI Key: MWWSFMDVAYGXBV-FGBJBKNOSA-N Synonym: doxorubicin hydrochloride,adriamycin,doxorubicin hcl,adriacin,adriblastina,rubex,adriamycin hydrochloride,adriblastin,adriamycin pfs PubChem CID: 129626538 IUPAC Name: (7R,9R)-7-[(2S,4R,5R,6R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride SMILES: [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O
Novus Biologicals™ Doxorubicin Hydrochloride Inhibitor
Low molecular weight, organic compound that may help regulate biological process.
Doxorubicin Hydrochloride, Thermo Scientific Chemicals PROMO
CAS: 25316-40-9 Molecular Formula: C27H30ClNO11 Molecular Weight (g/mol): 579.98 g/mol MDL Number: MFCD00077757,MFCD00077757,MFCD00941448 InChI Key: MWWSFMDVAYGXBV-FGBJBKNOSA-N Synonym: doxorubicin hydrochloride,adriamycin,doxorubicin hcl,adriacin,adriblastina,rubex,adriamycin hydrochloride,adriblastin,adriamycin pfs PubChem CID: 129626538 IUPAC Name: hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride SMILES: [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O
Thermo Scientific Chemicals Doxorubicin hydrochloride, 10 mg/ml in distilled water, sterile filtered
CAS: 25316-40-9 Molecular Formula: C27H30ClNO11 Molecular Weight (g/mol): 579.98 MDL Number: MFCD00077757,MFCD00077757,MFCD00941448 InChI Key: MWWSFMDVAYGXBV-FGBJBKNOSA-N PubChem CID: 129626538 IUPAC Name: hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride SMILES: [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O