Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxorubicin Hydrochloride, Fisher BioReagents

Catalog No. BP251610 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP251610 10 mg Amber Glass
BP251650 50 mg Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BP251610
This item is not returnable. View return policy

Doxorubicin Hydrochloride

Doxorubicin HCL is a chemotherapeutic agent. Antitumor, immunosuppressive, and antibiotic agent. It blocks RNA polymerase and reverse transcriptase. Also inhibits nucleic acid synthesis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25316-40-9
Molecular Formula C27H30ClNO11
Molecular Weight (g/mol) 579.98
MDL Number MFCD00077757,MFCD00077757,MFCD00941448
InChI Key MWWSFMDVAYGXBV-FGBJBKNOSA-N
Synonym doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs
PubChem CID 129626538
IUPAC Name (7R,9R)-7-[(2S,4R,5R,6R)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-9-(2-hydroxyacetyl)-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
SMILES [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O

Specifications

Decomposition Information 216°C
Melting Point 216°C
Color Orange
Assay Percent Range (by HPLC): ≥95%
Packaging Amber Glass
Quantity 10 mg
Merck Index 15, 3487
Solubility Information Soluble in water
Formula Weight 580
Physical Form Powder or Solid
Chemical Name or Material Doxorubicin Hydrochloride
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
May cause cancer. Risk of serious damage to eyes. Irritating to respiratory system and skin. May cause heritable genetic damage. Possible risk of impaired fertility. Possible risk of harm to the unborn child. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2
Flammability:1
Instability:0
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.