Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxorubicin hydrochloride, MP Biomedicals™

Catalog No. ICN15910101 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15910101 1 mg
ICN15910105 5 mg
ICN15910110 10 mg
ICN15910150 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15910101 Supplier MP Biomedicals, Inc Supplier No. 0215910101
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25316-40-9
Molecular Formula C27H30ClNO11
Molecular Weight (g/mol) 579.98
MDL Number MFCD00077757,MFCD00077757,MFCD00941448
InChI Key MWWSFMDVAYGXBV-FGBJBKNOSA-N
Synonym doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs
PubChem CID 129626538
IUPAC Name hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride
SMILES [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O

Specifications

Quantity 1 mg
Formula Weight 579.99
Chemical Name or Material Doxorubicin hydrochloride
EINECSNumber 246-818-3
Recommended Storage Store at Room Temperature (15° to 30°C)
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.