Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxycycline hyclate

Catalog No. AAJ6057922
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g

Doxycycline hyclate, Doryx, CAS # 24390-14-5, is the hyclate salt form of the broad-spectrum antibiotic known as doxycycline. | CAS: 24390-14-5 | C26H41ClN2O12 | 609.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General description
• Doxycycline hyclate is a broad-spectrum bacteriostatic antibiotic of the tetracycline class
• Doxycycline inhibits bacterial growth by preventing polypeptide formation via inhibition of at least one ribosomal subunit
• Known to reversibly bind the 30S ribosomal subunit and inhibit formation of the aminoacyl-tRNA/mRNA-ribosome complex
• Inhibits matrix metalloproteinases in many microorganisms
Applications
• Effective against the bacteria that cause Lyme disease, chlamydia, rickettsia and mycoplasma; suitable for antibiotic resistance testing and study of cellular mechanisms
• Intended for laboratory use only

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24390-14-5
Molecular Formula C26H41ClN2O12
Molecular Weight (g/mol) 609.066
MDL Number MFCD07357237
InChI Key ISZOFWGVNMFENH-YDLUHMIOSA-N
PubChem CID 131676157
IUPAC Name (4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;ethanol;dihydrate;hydrochloride
SMILES CCO.CCO.CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O.O.Cl

Specifications

Type Doxycycline Hyclate
Physical Form Powder
Color Yellow
Quantity 100 g
Merck Index 14,3440
Synonym Hydramycin; Doxycycline Hydrochloride Hemiethanolate Hemihydrate
Solubility Information Soluble in water to 50mg/ml; May require warming;Soluble in water,methanol; Sparingly soluble in ethanol; Insoluble in chloroform and ether.
Formula Weight 512.94 (503.94 Anhydrous)
Chemical Name or Material Doxycycline hyclate
missing translation for 'healthHazard1' H302-H315-H319-H335-H361fd
missing translation for 'healthHazard2' GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
RTECSNumber QI8925000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.