Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DP1 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299039PE
Description
DP1 Polyclonal antibody specifically detects DP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
DP1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
DP1E2F dimerization partner 1, DRTF1Dp-1, DRTF1-polypeptide 1, E2F-related transcription factor, transcription factor Dp-1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human DP1. | |
0.1 mL | |
Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing | |
7027 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction