Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DP1 Antibody (SR1878), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32227850UL
Description
DP1 Monoclonal antibody specifically detects DP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
DP1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry | |
SR1878 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
DP1E2F dimerization partner 1, DRTF1Dp-1, DRTF1-polypeptide 1, E2F-related transcription factor, transcription factor Dp-1 | |
A synthesized peptide derived from human DP1 (Uniprot #: Q14186) | |
50 μg | |
Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing | |
7027 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction