Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DP1 Monoclonal Antibody (1DP06 (TFD10)), Invitrogen™

Catalog No. PIMA511268
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA511268 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA511268 Supplier Thermo Scientific Supplier No. MA511268

Mouse Monoclonal Antibody

MA5-11268 targets DP-1 Transcription Factor in WB applications and shows reactivity with Human, mouse, and Rat samples. The MA5-11268 immunogen is recombinant DP-1 protein.

DP family contains at least two members, DP1 and 2. Structurally, DP1 and 2 are relatively conserved, yet their expression appears to be tissue specific. E2F 1, a functional target for the tumor suppressor protein Rb, heterodimerizes with DP1 and forms an active E2F transcriptional complex. The interaction between members of the E2F and the DP families is mediated, in part, by a leucine repeat which is weakly conserved between the two families.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1DP06 (TFD10)
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene TFDP1
Gene Accession No. Q08639, Q14186
Gene Alias AS1; ASRT1; cell cycle regulatory transcription factor DP1; DILC; down-regulated in liver cancer stem cells; DP; DP1; Dp-1; DRTF1; DRTF1-polypeptide 1; E2F dimerization partner 1; E2F-related transcription factor; MGC49004; OTTHUMP00000018767; Tfdp1; transcription factor Dp 1; transcription factor Dp-1
Gene Symbols LOC100360176, TFDP1
Host Species Mouse
Immunogen Recombinant DP-1 protein
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100360176, 21781, 361178, 7027
Target Species Human, Mouse, Rat
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.