Learn More
Invitrogen™ DP1 Recombinant Rabbit Monoclonal Antibody (8C0O3)

Description
Immunogen sequence: SGSCSAEDLK MARSLVPKAL EPYVTEMAQG TVGGVFITTA GSTSNGTRFS ASDLTNGADG MLATSSNGSQ YSGSRVETPV SYVGEDDEED DDFNENDEDD.
Specifications
Specifications
| Antigen | DP1 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 8C0O3 |
| Concentration | 0.35 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TFDP1 |
| Gene Accession No. | Q14186 |
| Gene Alias | AS1; ASRT1; cell cycle regulatory transcription factor DP1; DILC; down-regulated in liver cancer stem cells; DP; DP1; Dp-1; DRTF1; DRTF1-polypeptide 1; E2F dimerization partner 1; E2F-related transcription factor; MGC49004; OTTHUMP00000018767; Tfdp1; transcription factor Dp 1; transcription factor Dp-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.