Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599165
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599165 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599165 Supplier Invitrogen™ Supplier No. PA599165
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TFDP1.

DP family contains at least two members, DP1 and 2. Structurally, DP1 and 2 are relatively conserved, yet their expression appears to be tissue specific. E2F 1, a functional target for the tumor suppressor protein Rb, heterodimerizes with DP1 and forms an active E2F transcriptional complex. The interaction between members of the E2F and the DP families is mediated, in part, by a leucine repeat which is weakly conserved between the two families.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TFDP1
Gene Accession No. Q14186
Gene Alias AS1; ASRT1; cell cycle regulatory transcription factor DP1; DILC; down-regulated in liver cancer stem cells; DP; DP1; Dp-1; DRTF1; DRTF1-polypeptide 1; E2F dimerization partner 1; E2F-related transcription factor; MGC49004; OTTHUMP00000018767; Tfdp1; transcription factor Dp 1; transcription factor Dp-1
Gene Symbols LOC100360176, TFDP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TFDP1(Accession Q14186), corresponding to amino acid residues A373-D410.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100360176, 361178, 7027
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.