Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DP1 Recombinant Rabbit Monoclonal Antibody (JE52-55)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA536166
Description
DP1 Recombinant Monoclonal Antibody for Western Blot, IHC (P), Flow
DP family contains at least two members, DP1 and 2. Structurally, DP1 and 2 are relatively conserved, yet their expression appears to be tissue specific. E2F 1, a functional target for the tumor suppressor protein Rb, heterodimerizes with DP1 and forms an active E2F transcriptional complex. The interaction between members of the E2F and the DP families is mediated, in part, by a leucine repeat which is weakly conserved between the two families.
Specifications
DP1 | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q08639, Q14186 | |
LOC100360176, TFDP1 | |
Recombinant protein within Human DP1 aa 301-410. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
JE52-55 | |
Unconjugated | |
TFDP1 | |
AS1; ASRT1; cell cycle regulatory transcription factor DP1; DILC; down-regulated in liver cancer stem cells; DP; DP1; Dp-1; DRTF1; DRTF1-polypeptide 1; E2F dimerization partner 1; E2F-related transcription factor; MGC49004; OTTHUMP00000018767; Tfdp1; transcription factor Dp 1; transcription factor Dp-1 | |
Rabbit | |
Protein A | |
RUO | |
100360176, 21781, 361178, 7027 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction