Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DP2 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299040AF350
Description
DP2 Polyclonal specifically detects DP2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DP2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
DKFZp313O196, DP2Dp-2, E2F dimerization partner 2, FLJ39131, transcription factor Dp-2, transcription factor Dp-2 (E2F dimerization partner 2) | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human DP2. | |
0.1 mL | |
Cell Cycle and Replication | |
7029 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction