Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPAGT1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | DPAGT1 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
DPAGT1 Polyclonal specifically detects DPAGT1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DPAGT1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ALG7, CDG1J, CDG-Ij, D11S366, DGPTDPAGT2UAGT, dolichyl-phosphate (UDP-N-acetylglucosamine)N-acetylglucosaminephosphotransferase 1 (GlcNAc-1-P tra, dolichyl-phosphate (UDP-N-acetylglucosamine)N-acetylglucosaminephosphotransferase 1 (GlcNAc-1-P transferase), dolichyl-phosphate alpha-N-acetylglucosaminyltransferase, EC 2.7.8, EC 2.7.8.15, G1PT, GlcNAc-1-P transferase, GPTDPAGT, N-acetylglucosamine-1-phosphate transferase, UDP-GlcNAc:dolichyl-phosphate N-acetylglucosaminephosphotransferase, UDP-N-acetylglucosamine--dolichyl-phosphateN-acetylglucosaminephosphotransferase, UGAT | |
DPAGT1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Human | |
Q9H3H5 | |
1798 | |
This antibody was developed against a recombinant protein corresponding to amino acids: GHFIAARLCGQDLNKTSRQQIPESQ | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title