Learn More
Invitrogen™ DPCD Monoclonal Antibody (OTI4B9)

Description
DPCD Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | DPCD |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI4B9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DPCD |
| Gene Accession No. | Q9BVM2 |
| Gene Alias | 5330431N19Rik; deleted in a mouse model of primary ciliary dyskinesia; deleted in primary ciliary dyskinesia; deleted in primary ciliary dyskinesia homolog (mouse); DPCD; Ndac; neighbor of Dac; protein DPCD; RGD1307648; RP11-529I10.4; RP11-529I104 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.