Learn More
Invitrogen™ DPCD Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114050
Description
Immunogen sequence: APLGAGNLGP ELIKESNANP IFMRKDTKMS FQWRIRNLPY PKDVYSVSVD For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
May play a role in the formation or function of ciliated cells.
Specifications
| DPCD | |
| Polyclonal | |
| Unconjugated | |
| DPCD | |
| 5330431N19Rik; deleted in a mouse model of primary ciliary dyskinesia; deleted in primary ciliary dyskinesia; deleted in primary ciliary dyskinesia homolog (mouse); DPCD; Ndac; neighbor of Dac; protein DPCD; RGD1307648; RP11-529I10.4; RP11-529I104 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25911 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BVM2 | |
| DPCD | |
| Synthetic peptide directed towards the middle region of human RP11-529I10.4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.