Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPCR-1 Antibody (213201), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3347PEATT594
Description
DPCR-1 Monoclonal antibody specifically detects DPCR-1 in Human samples. It is validated for Flow CytometrySpecifications
DPCR-1 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
135656 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Flow Cytometry | |
213201 | |
Flow Cytometry | |
bCX105N19.6, C6orf37, diffuse panbronchiolitis critical region 1, MGC126710, MGC126712, PBLTdiffuse panbronchiolitis critical region 1 protein | |
NS0 mouse myeloma cell line transfected with human DPCR-1, Met1-Arg235, Accession # AAH69477 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction