Learn More
Invitrogen™ DPCR1 Monoclonal Antibody (213201)

Description
This antibody stains human DPCR-1-transfected cells but not irrelevant transfectants. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Specifications
Specifications
| Antigen | DPCR1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 213201 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | DPCR1 |
| Gene Accession No. | Q3MIW9 |
| Gene Alias | bCX105N19.6; C6orf37; diffuse panbronchiolitis critical region 1; diffuse panbronchiolitis critical region protein 1; DPCR1; MGC126710; MGC126712; Mucin-like protein 3; MUCL3; PBLT |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.