Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPCR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553115
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553115 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553115 Supplier Invitrogen™ Supplier No. PA553115
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KTTSTTEKTT RTPEKPTLYS EKTICTKGKN TPVPEKPTEN LGNTTLTTET IKAPVKSTEN PEKTAAVTKT IKPSVKVTGD KSLTTTSSHL NKTEVTHQVP TGSFTLITSR TKLSSITSEA TGNESHPYLN KDGSQK Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 33%.

The DPCR1 gene, known as diffuse panbronchiolitis critical region 1, encodes a protein involved in immune responses and epithelial cell function, particularly in the respiratory system. Although the specific functions of DPCR1 are not extensively characterized, the gene is located within a region linked to diffuse panbronchiolitis, a chronic inflammatory lung disease. DPCR1 is thought to contribute to immune regulation, potentially influencing innate immune responses and epithelial cell interactions critical for maintaining respiratory health. Its expression in airway epithelial cells suggests a role in modulating mucus production, barrier integrity, and protection against pathogens. Aberrant expression of DPCR1 may impact respiratory function, contributing to conditions such as asthma or chronic obstructive pulmonary disease (COPD) by affecting immune responses and inflammation. Understanding DPCR1's functions and regulatory mechanisms in the respiratory system is essential for exploring its potential as a therapeutic target for respiratory diseases characterized by chronic inflammation and immune dysregulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPCR1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DPCR1
Gene Accession No. Q3MIW9
Gene Alias bCX105N19.6; C6orf37; diffuse panbronchiolitis critical region 1; diffuse panbronchiolitis critical region protein 1; DPCR1; MGC126710; MGC126712; Mucin-like protein 3; MUCL3; PBLT
Gene Symbols DPCR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DPCR1. Recombinant protein control fragment (Product #RP-90421).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135656
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.