Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DPCR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543536
Description
Peptide sequence: SHLNKTEVTH QVPTGSFTLI TSRTKLSSIT SEATGNESHP YLNKDGSQKG Sequence homology: Human: 100%.
The DPCR1 gene, known as diffuse panbronchiolitis critical region 1, encodes a protein involved in immune responses and epithelial cell function, particularly in the respiratory system. Although the specific functions of DPCR1 are not extensively characterized, the gene is located within a region linked to diffuse panbronchiolitis, a chronic inflammatory lung disease. DPCR1 is thought to contribute to immune regulation, potentially influencing innate immune responses and epithelial cell interactions critical for maintaining respiratory health. Its expression in airway epithelial cells suggests a role in modulating mucus production, barrier integrity, and protection against pathogens. Aberrant expression of DPCR1 may impact respiratory function, contributing to conditions such as asthma or chronic obstructive pulmonary disease (COPD) by affecting immune responses and inflammation. Understanding DPCR1's functions and regulatory mechanisms in the respiratory system is essential for exploring its potential as a therapeutic target for respiratory diseases characterized by chronic inflammation and immune dysregulation.
Specifications
DPCR1 | |
Polyclonal | |
Unconjugated | |
DPCR1 | |
bCX105N19.6; C6orf37; diffuse panbronchiolitis critical region 1; diffuse panbronchiolitis critical region protein 1; DPCR1; MGC126710; MGC126712; Mucin-like protein 3; MUCL3; PBLT | |
Rabbit | |
Affinity Chromatography | |
RUO | |
135656 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q3MIW9 | |
DPCR1 | |
Synthetic peptide directed towards the C-terminal of human DPCR1 (aa 383-432). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction