Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPEP2 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300197DL594
Description
DPEP2 Polyclonal specifically detects DPEP2 in Human samples. It is validated for ELISA.Specifications
DPEP2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
64174 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
DyLight 594 | |
dipeptidase 2, EC 3.4.13.19, MBD2 | |
Produced in rabbits immunized with purified, recombinant DPEP2 (AAH24021.1; Met1-Ser376). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction