Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPEP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100487
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100487 Supplier Invitrogen™ Supplier No. PA5100487
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DPEP2.

Probable metalloprotease which hydrolyzes leukotriene D4 (LTD4) into leukotriene E4 (LTE4).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPEP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DPEP2
Gene Accession No. Q5M872, Q8C255, Q9H4A9
Gene Alias dipeptidase 2; DPEP2; F630103D06Rik; MBD2; MBD-2; Membrane-bound dipeptidase 2; putative membrane-bound dipeptidase 2; UNQ284/PRO323
Gene Symbols DPEP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human DPEP2(Accession Q9H4A9), corresponding to amino acid residues L119-V169.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291984, 319446, 64174
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.