Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DPEP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572282
Description
DPEP2 Polyclonal Antibody for Western Blot
Probable metalloprotease which hydrolyzes leukotriene D4 (LTD4) into leukotriene E4 (LTE4).
Specifications
DPEP2 | |
Polyclonal | |
Unconjugated | |
DPEP2 | |
dipeptidase 2; DPEP2; F630103D06Rik; MBD2; MBD-2; Membrane-bound dipeptidase 2; putative membrane-bound dipeptidase 2; UNQ284/PRO323 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
319446, 64174 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8C255, Q9H4A9 | |
DPEP2 | |
KLH conjugated synthetic peptide between 220-254 amino acids from the Central region of human DPEP2. | |
200 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction