Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPEP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563597
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563597 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563597 Supplier Invitrogen™ Supplier No. PA563597
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVREESRAQS PVEAEFPYGQ LSTSCHSHLV PQNGHQATHL EVTKQPTNRV Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 56%.

This gene encodes a membrane-bound glycoprotein from the family of dipeptidases involved in hydrolytic metabolism of various dipeptides, including penem and carbapenem beta-lactam antibiotics. This gene is located on chromosome 16 in a cluster with another member of this family. Alternatively spliced transcript variants that encode different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPEP3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DPEP3
Gene Accession No. Q9H4B8
Gene Alias 1700018F16Rik; AI507039; Dipeptidase 3; DPEP3; Mbd3; MBD-3; Membrane-bound dipeptidase 3; metallopeptidase (family M19); MLZ-136; Protein expressed in male leptotene and zygotene spermatocytes 136; putative membrane-bound dipeptidase 3; testis secretory sperm-binding protein Li 211a; UNQ834/PRO1772
Gene Symbols DPEP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DPEP3. Recombinant protein control fragment (Product #RP-101813).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64180
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.