Learn More
Invitrogen™ DPF2 Polyclonal Antibody

Description
Recommended positive controls: DPF2 tansfected 293T cell. Predicted reactivity: Mouse (98%), Rat (99%), Xenopus laevis (84%), Chicken (90%), Chimpanzee (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | DPF2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.67 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Dpf2 |
| Gene Accession No. | Q61103, Q92785 |
| Gene Alias | 2210010M07Rik; apoptosis response zinc finger protein; BAF45D; BRG1-associated factor 45D; D4, zinc and double PHD fingers family 2; double PHD fingers 2; Dpf2; neuro-d4/requiem; protein requiem; Req; requiem; requiem, apoptosis response zinc finger; UBID4; ubi-d4; Zinc finger protein ubi-d4 |
| Gene Symbols | Dpf2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.