Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPF2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501565566
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-5566 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-5566 Supplier Bethyl Laboratories, Inc Supplier No. A303596A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

Requiem is a member of the d4 domain family, characterized by a zinc finger-like structural motif. This protein functions as a transcription factor which is necessary for the apoptotic response following deprivation of survival factors. It is expressed in numerous tissues in both cytosolic and nuclear regions and likely serves a regulatory role in rapid hematopoietic cell growth and turnover. Requiem has been found to act as an adaptor protein that links the NF-kappa-B and SWI/SNF chromatin remodeling factor and is considered a candidate gene for multiple endocrine neoplasia type I, an inherited cancer syndrome involving multiple parathyroid, enteropancreatic, and pituitary tumors.

Specifications

Antigen DPF2
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene DPF2
Gene Accession No. Q92785
Gene Alias REQ, UBID4, ubi-d4; BAF45D; BRG1-associated factor 45D; apoptosis response zinc finger protein; protein requiem; requiem, apoptosis response zinc finger; zinc finger protein ubi-d4
Gene Symbols DPF2
Host Species Rabbit
Immunogen Between 341 and 391
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5977
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.