Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPF2 Rabbit anti-Human, Rat, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABE132725UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1327-25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1327-25UG Supplier MilliporeSigma Supplier No. ABE132725UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect BRG1-associated factor 45D using this rabbit polyclonal Anti-DPF2 validated for use in Western Blotting and Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPF2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols DPF2,BAF45D,REQ,UBID4
Host Species Rabbit
Immunogen KLH-conjugated liner peptide that corresponding to 17 amino acids from the N-terminal region of human BRG1-associated factor 45D (DPF2).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects BRG1-associated factor 45D in human, mouse, and rat species. It targets an epitope within 17 amino acids from the N-terminal half. The immunogen sequence is conserved in both isoforms.
Target Species Human, Rat, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.