Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPF3 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282087IR
This item is not returnable.
View return policy
Description
DPF3 Polyclonal antibody specifically detects DPF3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
DPF3 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
8110 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
CERD4, D4, zinc and double PHD fingers, family 3 | |
DPF3 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human DPF3. The immunogen is located within amino acids 220 - 270 of DPF3. Amino Acid Squence: SPPNHRNENHRPQKGPDG | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction