Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DPF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531963
Description
Recommended positive controls: IMR32. Predicted reactivity: Mouse (99%), Rat (100%), Xenopus laevis (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the D4 protein family. The encoded protein is a transcription regulator that binds acetylated histones and is a component of the BAF chromatin remodeling complex. Alternate splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Jul 2013].
Specifications
| DPF3 | |
| Polyclonal | |
| Unconjugated | |
| DPF3 | |
| 2810403B03Rik; 6530402L11Rik; BAF45C; BRG1-associated factor 45C; C78788; CERD4; cer-d4; cerebellum D4; D4 zinc and double PHD finger family 3; D4, zinc and double PHD fingers, family 3; double PHD fingers 3; Dpf3; Zinc finger protein cer-d4; zinc finger protein DPF3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 70127, 8110 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P58269, Q92784 | |
| DPF3 | |
| Recombinant protein encompassing a sequence within the center region of human CERD4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction