Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPF3 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA584737
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584737 Supplier Thermo Scientific Supplier No. PA584737

Rabbit Polyclonal Antibody

Immunogen sequence: KLRLLEIKPE VELPLKKDGF TSESTTLEAL LRGEGVEKKV DAREEESIQE IQRVLENDEN VEEGNEEEDL EEDIPKRKNR TRGRA

This gene encodes a member of the D4 protein family. The encoded protein is a transcription regulator that binds acetylated histones and is a component of the BAF chromatin remodeling complex. Alternate splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Jul 2013].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPF3
Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide, pH 7.2
Gene DPF3
Gene Accession No. Q92784
Gene Alias 2810403B03Rik, 6530402L11Rik, BAF45C, BRG1-associated factor 45C, C78788, CERD4, cer-d4, cerebellum D4, D4 zinc and double PHD finger family 3, D4, zinc and double PHD fingers, family 3, double PHD fingers 3, Dpf3, Zinc finger protein cer-d4, zinc finger protein DPF3
Gene Symbols DPF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DPF3
Purification Method Anitgen affinity chromatography, Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8110
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.