Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPH2 Antibody (OTI3A12), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270587H
Description
DPH2 Monoclonal specifically detects DPH2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
DPH2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
DPH2 | |
Full length recombinant protein of human DPH2 (NP_001375) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI3A12 | |
Western Blot, Immunohistochemistry | |
diphthamide biosynthesis protein 2, Diphthamide biosynthesis protein 2 homolog-like 2, diptheria toxin resistance protein required for diphthamide biosynthesis-like 2, diptheria toxin resistance protein required for diphthamide biosynthesis-like 2(S. cerevisiae), DPH2 homolog, DPH2 homolog (S. cerevisiae), DPH2L2diphthamide biosynthesis-like protein 2, DPH2-like 2, DPH2-like 2 (S. cerevisiae), HsDph2 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
1802.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction