Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557215
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557215 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557215 Supplier Invitrogen™ Supplier No. PA557215
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDFQYDEDSE TYFYPCPCGD NFSITKEDLE NGEDVATCPS CSLIIKVIYD KDQFVCGETV PAPSANKELV K Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen DPH3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DPH3
Gene Accession No. Q96FX2
Gene Alias CSL-type zinc finger-containing protein 2; deafness locus putative guanine nucleotide exchange factor interacting protein 1; Del-GEF interacting protein; delGEF-interacting protein 1; DELGIP; DelGIP1; DESR1; diphthamide biosynthesis 3; diphtheria toxin and Pseudomonas exotoxin A sensitivity required gene 1; DPH3; DPH3 homolog; DPH3 homolog (KTI11, S. cerevisiae); DPH3, KTI11 homolog; DPH3A; DPH3A, KTI11 homolog A; KTI11; ZCSL2; zinc finger, CSL domain containing 2; zinc finger, CSL-type containing 2
Gene Symbols DPH3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DPH3. Recombinant protein control fragment (Product #RP-96377).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285381
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.