Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPH5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561242
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561242 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561242 Supplier Invitrogen™ Supplier No. PA561242
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVNQAAQQLL EIVQNQRIRG EEPAVTEETL CVGLARVGAD DQKIAAGTLR QMCTVDLGEP LHSLIITGGS IHPMEMEMLS LFSIPENSSE SQSING Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.

This gene encodes a component of the diphthamide synthesis pathway. Diphthamide is a post-translationally modified histidine residue found only on translation elongation factor 2. It is conserved from archaebacteria to humans, and is targeted by diphtheria toxin and Pseudomonas exotoxin A to halt cellular protein synthesis. The yeast and Chinese hamster homologs of this protein catalyze the trimethylation of the histidine residue on elongation factor 2, resulting in a diphthine moiety that is subsequently amidated to yield diphthamide. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPH5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DPH5
Gene Accession No. Q9H2P9
Gene Alias AD-018; CGI-30; diphthamide biosynthesis 5; diphthamide biosynthesis methyltransferase; Diphthine methyl ester synthase; diphthine synthase; DPH5; DPH5 homolog; HSPC143; NPD015; protein x 0011
Gene Symbols DPH5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DPH5. Recombinant protein control fragment (Product #RP-99786).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51611
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.