Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519027
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519027 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519027 Supplier Invitrogen™ Supplier No. PA519027
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Dolichol-phosphate mannose (Dol-P-Man) serves as a donor of mannosyl residues on the lumenal side of the endoplasmic reticulum (ER). Lack of Dol-P-Man results in defective surface expression of GPI-anchored proteins. Dol-P-Man is synthesized from GDP-mannose and dolichol-phosphate on the cytosolic side of the ER by the enzyme dolichyl-phosphate mannosyltransferase. Human DPM1 lacks a carboxy-terminal transmembrane domain and signal sequence and is regulated by DPM2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPM1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene DPM1
Gene Accession No. O60762
Gene Alias AI118379; AI194292; CDGIE; dolichol monophosphate mannose synthase; dolichol-phosphate (beta-D) mannosyltransferase 1; Dolichol-phosphate mannose synthase; Dolichol-phosphate mannose synthase subunit 1; Dolichol-phosphate mannosyltransferase; Dolichol-phosphate mannosyltransferase subunit 1; dolichyl-phosphate beta-D-mannosyltransferase; Dolichyl-phosphate beta-D-mannosyltransferase subunit 1; dolichyl-phosphate mannosyltransferase 1; dolichyl-phosphate mannosyltransferase polypeptide 1 catalytic subunit; dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit; DPM synthase; DPM synthase complex, catalytic subunit; DPM synthase subunit 1; Dpm1; Mannose-P-dolichol synthase; Mannose-P-dolichol synthase subunit 1; MPD synthase; MPD synthase subunit 1; MPDS; OTTMUSP00000017164; P9705.3; RP23-391M18.1; SED3; YPR183W
Gene Symbols DPM1
Host Species Goat
Immunogen Synthetic peptide sequence (RELEVRSPRQNKYS) corresponding to the N-terminus amino acids of DPM1 (aa 15-29).
Purification Method Antigen affinity chromatography, Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8813
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.