Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ DPO-1
GREENER_CHOICESDP

Catalog No. 253350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-335-0 50 mg
25-331-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-335-0 Supplier R&D Systems™ Supplier No. 2533/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Blocker of KV1.5 channel and IKur current

Blocker of IKur ultrarapid delayed rectifier potassium current and KV1.5 channels (IC50 = 0.31 μM for rKV1.5). Displays selectivity for inhibition of IKur over Ito (8-fold), IK1, IKr and IKs (20-fold) in native myocytes and selectivity for rat recombinant KV1.5 over KV3.1 (∽ 15-fold). Increases action potential duration in atrial but not ventricular myocytes and prevents atrial arrhythmia.

Chemical Identifiers

CAS 43077-30-1
Molecular Formula C22H29OP

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 340.44
Purity >99%
Molecular Weight (g/mol) 340.45
Chemical Name or Material DPO-1
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.