Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chembio Diagnostic Systems, Inc.™ DPP HIV-Syphilis PROMO

Click to view available options
Includes:
20 Kits and 1 Reader
35 Kits and 2 Readers
Description
- Test utilizes patented Dual Path Platform (DPP) technology, providing results in 15 minutes
- Easy to use and can be stored at room temperature
- Single-use rapid, qualitative, multiplex, immunoassay for the detection of antibodies to HIV-1/2, and/or Treponema pallidum bacteria (the causative agent of syphilis)
- Patented DPP technology allows for higher sensitivity: >99% for HIV and >94% for T. pallidum
- Only requires 10 μL of either fingerstick whole blood, venous whole blood or plasma
- Provides results in 15 minutes and is read with the DPP Micro Reader
Specifications
Specifications
Content And Storage | 2°C to 25°C |
CE Marker | Yes |
Clia Complexity | Waived |
Detectable Analytes | HIV, T. pallidum |
Sample Type | Fingerstick Whole Blood, Venous Whole Blood, Plasma |
Type | Rapid Test |
For Use With (Application) | DPP Rapid Test |
Includes | 20 Kits and 1 Reader |
Packaging Type | Kit |
Quantity | 20 Tests/Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction