Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPP10 Antibody (004), DyLight 350 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289978UV
Description
DPP10 Monoclonal antibody specifically detects DPP10 in Human samples. It is validated for ELISASpecifications
DPP10 | |
Monoclonal | |
DyLight 350 | |
dipeptidyl peptidase 10, Dipeptidyl peptidase IV-related protein 3, dipeptidyl peptidase like protein 2, Dipeptidyl peptidase X, Dipeptidyl peptidase-like protein 2, dipeptidyl-peptidase 10, dipeptidyl-peptidase 10 (non-functional), DPL2, DPP X, DPPY, DPRP-3, DPRP3dipeptidylpeptidase 10, inactive dipeptidyl peptidase 10, KIAA1492 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human DPP10 (Q8N608-1; Leu 56-Glu 796). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
004 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
57628 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction