Learn More
DPP10 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5111885
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: CSRKYEMTSD TWISK Homology: Mouse - 14/15 amino acid residues identical; human - 12/15 amino acid residues identical.
A novel gene, DPP10, encodes a homolog of dipeptidyl peptidases (DPPs) that cleave terminal dipeptides from cytokines and chemokines, and it presents a potential new target for asthma therapy. The DPP10 protein shares features with members of the S9B family of DPP serine proteases, which includes DPP4, a widely expressed enzyme that plays a central role in chemokine processing as part of the innate immune system. The locus displays a complex pattern of transcript splicing, with eight alternate first axons; four of which localize within the LD block strongly associated with asthma.
Specifications
DPP10 (extracellular) | |
Polyclonal | |
Unconjugated | |
DPP10 | |
6430601K09Rik; Dipeptidyl peptidase IV-related protein 3; Dipeptidyl peptidase X; Dipeptidyl peptidase-like protein 2; DPL2; DPP X; DPP10; DPPY; DPRP3; DPRP-3; HGNC:20823; Inactive dipeptidyl peptidase 10; KIAA1492; Kv4 potassium channel auxiliary subunit; OTTHUMP00000203618 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
269109, 363972, 57628 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
Q6NXK7, Q6Q629, Q8N608 | |
DPP10 | |
Peptide corresponding to amino acid residues 356-370 of rat DPP10 (Extracellular, C-terminus) | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.