Learn More
Invitrogen™ DPP10 Monoclonal Antibody (OTI2A5)

Description
DPP10 Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | DPP10 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2A5 |
| Concentration | 0.43 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DPP10 |
| Gene Accession No. | Q8N608 |
| Gene Alias | 6430601K09Rik; dipeptidyl peptidase 10; dipeptidyl peptidase IV-related protein 3; dipeptidyl peptidase like 10; dipeptidyl peptidase X; Dipeptidyl peptidase-like protein 2; dipeptidylpeptidase 10; dipeptidyl-peptidase 10 (inactive); dipeptidyl-peptidase 10 (non-functional); DPL2; DPP X; DPP10; DPPY; Dprp3; DPRP-3; HGNC:20823; inactive dipeptidyl peptidase 10; KIAA1492; Kv4 potassium channel auxiliary subunit; OTTHUMP00000203618 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.