Learn More
DPP10 Mouse anti-Human, Clone: OTI1A5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503585

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
A novel gene, DPP10, encodes a homolog of dipeptidyl peptidases (DPPs) that cleave terminal dipeptides from cytokines and chemokines, and it presents a potential new target for asthma therapy. The DPP10 protein shares features with members of the S9B family of DPP serine proteases, which includes DPP4, a widely expressed enzyme that plays a central role in chemokine processing as part of the innate immune system. The locus displays a complex pattern of transcript splicing, with eight alternate first axons; four of which localize within the LD block strongly associated with asthma.Specifications
DPP10 | |
Monoclonal | |
Unconjugated | |
DPP10 | |
DPL2, DPPY, DPRP-3, DPRP3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
57628 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1A5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8N608 | |
DPP10 | |
Full length human recombit protein of human DPP10 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.