Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPP10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109655
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109655 Supplier Invitrogen™ Supplier No. PA5109655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GYIASMILKS DEKLFKCGSV VAPITDLKLY ASAFSERYLG MPSKEESTYQ AASVLHNVHG LKEENILIIH GTADTKVHFQ HSAELIKHLI KAGVNYTMQV Y.

A novel gene, DPP10, encodes a homolog of dipeptidyl peptidases (DPPs) that cleave terminal dipeptides from cytokines and chemokines, and it presents a potential new target for asthma therapy. The DPP10 protein shares features with members of the S9B family of DPP serine proteases, which includes DPP4, a widely expressed enzyme that plays a central role in chemokine processing as part of the innate immune system. The locus displays a complex pattern of transcript splicing, with eight alternate first axons; four of which localize within the LD block strongly associated with asthma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPP10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DPP10
Gene Accession No. Q8N608
Gene Alias 6430601K09Rik; dipeptidyl peptidase 10; dipeptidyl peptidase IV-related protein 3; dipeptidyl peptidase like 10; dipeptidyl peptidase X; Dipeptidyl peptidase-like protein 2; dipeptidylpeptidase 10; dipeptidyl-peptidase 10 (inactive); dipeptidyl-peptidase 10 (non-functional); DPL2; DPP X; DPP10; DPPY; Dprp3; DPRP-3; HGNC:20823; inactive dipeptidyl peptidase 10; KIAA1492; Kv4 potassium channel auxiliary subunit; OTTHUMP00000203618
Gene Symbols DPP10
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 400-500 of human DPP10 (NP_001308842.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57628
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.