Learn More
Invitrogen™ DPP10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533471
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Monkey, Marmoset, Panda, Bovine, Dog, Horse, Guinea pig (100%); Galago, Bat, Rabbit, Pig (94%); Opossum, Turkey, Zebra finch, Chicken, Xenopus (88%); Mouse, Rat, Hamster, Elephant (82%). Proteinase K is required for antigen retrieval.
A novel gene, DPP10, encodes a homolog of dipeptidyl peptidases (DPPs) that cleave terminal dipeptides from cytokines and chemokines, and it presents a potential new target for asthma therapy. The DPP10 protein shares features with members of the S9B family of DPP serine proteases, which includes DPP4, a widely expressed enzyme that plays a central role in chemokine processing as part of the innate immune system. The locus displays a complex pattern of transcript splicing, with eight alternate first axons; four of which localize within the LD block strongly associated with asthma.
Specifications
| DPP10 | |
| Polyclonal | |
| Unconjugated | |
| DPP10 | |
| 6430601K09Rik; dipeptidyl peptidase 10; dipeptidyl peptidase IV-related protein 3; dipeptidyl peptidase like 10; dipeptidyl peptidase X; Dipeptidyl peptidase-like protein 2; dipeptidylpeptidase 10; dipeptidyl-peptidase 10 (inactive); dipeptidyl-peptidase 10 (non-functional); DPL2; DPP X; DPP10; DPPY; Dprp3; DPRP-3; HGNC:20823; inactive dipeptidyl peptidase 10; KIAA1492; Kv4 potassium channel auxiliary subunit; OTTHUMP00000203618 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100053255, 100714259, 476121, 57628, 617222 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q8N608 | |
| DPP10 | |
| Synthetic 17 amino acid peptide from internal region of human DPP10. | |
| 50 μg | |
| Primary | |
| Human, Canine, Monkey, Bovine, Horse, Guinea Pig | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.