Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPP2 Monoclonal Antibody (2G1C10A9)
GREENER_CHOICE

Catalog No. PIMA530490
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530490 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530490 Supplier Invitrogen™ Supplier No. MA530490
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human DPPII.

The protein encoded by this gene is a post-proline cleaving aminopeptidase expressed in quiescent lymphocytes. The resting lymphocytes are maintained through suppression of apoptosis, a state which is disrupted by inhibition of this novel serine protease. The enzyme has strong sequence homology with prolylcarboxypeptidase and is active at both acidic and neutral pH.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPP2
Applications ELISA
Classification Monoclonal
Clone 2G1C10A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene DPP7
Gene Accession No. Q9UHL4
Gene Alias carboxytripeptidase; dipeptidyl aminopeptidase II; dipeptidyl arylamidase II; dipeptidyl peptidase 2; Dipeptidyl peptidase 7; dipeptidyl peptidase II; dipeptidyl-peptidase 2; dipeptidylpeptidase 7; dipeptidyl-peptidase II; DPP II; DPP2; Dpp7; DPPII; QPP; Quiescent cell proline dipeptidase
Gene Symbols DPP7
Host Species Mouse
Immunogen Human cell-derived DPPII protein (Met1-Leu492).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29952
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.