Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547489
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547489 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547489 Supplier Invitrogen™ Supplier No. PA547489
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, approximately 60% cross-reactivity with recombinant mouse DPPII is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

The protein encoded by this gene is a post-proline cleaving aminopeptidase expressed in quiescent lymphocytes. The resting lymphocytes are maintained through suppression of apoptosis, a state which is disrupted by inhibition of this novel serine protease. The enzyme has strong sequence homology with prolylcarboxypeptidase and is active at both acidic and neutral pH.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPP2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene DPP7
Gene Accession No. Q9UHL4
Gene Alias carboxytripeptidase; dipeptidyl aminopeptidase II; dipeptidyl arylamidase II; dipeptidyl peptidase 2; Dipeptidyl peptidase 7; dipeptidyl peptidase II; dipeptidyl-peptidase 2; dipeptidylpeptidase 7; dipeptidyl-peptidase II; DPP II; DPP2; Dpp7; DPPII; QPP; Quiescent cell proline dipeptidase
Gene Symbols DPP7
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human DPPII/QPP/DPP7 Gly22-Leu492.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29952
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.