Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554369
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554369 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554369 Supplier Invitrogen™ Supplier No. PA554369
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MFARNAFTVL AMMDYPYPTD FLGPLPANPV KVGCDRLLSE AQRITGLRAL AGLVYNASGS EHCYDIYRLY H Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 83%.

The protein encoded by this gene is a post-proline cleaving aminopeptidase expressed in quiescent lymphocytes. The resting lymphocytes are maintained through suppression of apoptosis, a state which is disrupted by inhibition of this novel serine protease. The enzyme has strong sequence homology with prolylcarboxypeptidase and is active at both acidic and neutral pH.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DPP7
Gene Accession No. Q9UHL4
Gene Alias carboxytripeptidase; dipeptidyl aminopeptidase II; dipeptidyl arylamidase II; dipeptidyl peptidase 2; Dipeptidyl peptidase 7; dipeptidyl peptidase II; dipeptidyl-peptidase 2; dipeptidylpeptidase 7; dipeptidyl-peptidase II; DPP II; DPP2; Dpp7; DPPII; QPP; Quiescent cell proline dipeptidase
Gene Symbols DPP7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DPP2. Recombinant protein control fragment (Product #RP-93071).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29952
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.