Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPP3 Antibody (SR2247), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | DPP3 |
---|---|
Clone | SR2247 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50 |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Description
DPP3 Monoclonal antibody specifically detects DPP3 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
DPP3 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Dipeptidyl aminopeptidase III, Dipeptidyl arylamidase III, dipeptidyl peptidase 3, Dipeptidyl peptidase III, dipeptidylpeptidase 3, dipeptidyl-peptidase 3, dipeptidylpeptidase III, DPP III, DPPIII, EC 3.4.14.4, FLJ11387, FLJ22331 | |
A synthesized peptide derived from human DPP3 (Uniprot #: Q9NY33) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2247 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
10072 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title