Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPP3 Monoclonal Antibody (OTI 2A8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
$231.00 - $516.29
Specifications
Antigen | DPP3 |
---|---|
Clone | OTI 2A8 |
Concentration | 0.66 mg/mL |
Applications | Flow Cytometry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-254-1000
![]() ![]() ![]() |
OriGene
TA503286S |
30 μL |
Each for $231.00
|
|
|||||
50-167-4220
![]() ![]() |
OriGene
TA503286 |
100 μL |
Each for $516.29
|
|
|||||
Description
DPP3 Monoclonal antibody specifically detects DPP3 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
DPP3 | |
0.66 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Q9NY33 | |
IgG2a | |
-20° C, Avoid Freeze/Thaw Cycles |
OTI 2A8 | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
10072 | |
Primary | |
DPP3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title