Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPP3 Mouse anti-Human, Biotin, Clone: OTI2D9, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA700273

Description
This gene encodes a protein that is a member of the M49 family of metallopeptidases. This cytoplasmic protein binds a single zinc ion with its zinc-binding motif (HELLGH) and has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Increased activity of this protein is associated with endometrial and ovarian cancers. Alternate transcriptional splice variants have been characterized.Specifications
DPP3 | |
OTI2D9 | |
Biotin | |
DPP3 | |
DPPIII | |
Mouse | |
Affinity Chromatography | |
RUO | |
10072 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Monoclonal | |
0.5 mg/mL | |
PBS with 10mg/ml BSA, 50% glycerol and 0.05% sodium azide | |
Q9NY33 | |
DPP3 | |
Full length human recombit protein of human DPP3 produced in HEK293T cell. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction