Learn More
DPP3 Mouse anti-Human, Clone: OTI2A12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503243

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein that is a member of the M49 family of metallopeptidases. This cytoplasmic protein binds a single zinc ion with its zinc-binding motif (HELLGH) and has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Increased activity of this protein is associated with endometrial and ovarian cancers. Alternate transcriptional splice variants have been characterized.Specifications
DPP3 | |
Monoclonal | |
Unconjugated | |
DPP3 | |
DPPIII | |
Mouse | |
Affinity Chromatography | |
RUO | |
10072 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2A12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NY33 | |
DPP3 | |
Full length human recombit protein of human DPP3 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.