Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPP4 Mouse anti-Human, Unconjugated, Clone: 134-2C2, Abnova™

Catalog No. 89337666 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-666 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-666 Supplier Abnova Corporation Supplier No. MAB21173
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

DPP4 Monoclonal antibody specifically detects DPP4 in Human samples. It is validated for IHC-Fr, IF, Flow Cyt.

Specifications

Antigen DPP4
Applications Immunohistochemistry (Frozen), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone 134-2C2
Conjugate Unconjugated
Description dipeptidyl-peptidase 4
Gene DPP4
Gene Alias ADABP, ADCP2, CD26, DPPIV, TP103
Host Species Mouse
Immunogen A synthetic peptide corresponding to terminal region of human DPP4
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1803
Target Species Human
Content And Storage Store at 4°C
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.