Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPP4 Mouse anti-Human, Unconjugated, Clone: 202.36, Abnova™

Catalog No. 89337662 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-662 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-662 Supplier Abnova Corporation Supplier No. MAB21169
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

DPP4 Monoclonal antibody specifically detects DPP4 in Human samples. It is validated for IHC-Fr, IF, Flow Cyt.

Specifications

Antigen DPP4
Applications Immunohistochemistry (Frozen), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone 202.36
Conjugate Unconjugated
Description dipeptidyl-peptidase 4
Gene DPP4
Gene Alias ADABP, ADCP2, CD26, DPPIV, TP103
Host Species Mouse
Immunogen Human T cell clone
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1803
Target Species Human
Content And Storage Store at 4°C
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.